Learn More
Waters Corp Check Valve Assembly, Spring Loaded, Primary, THF
Catalog No. 502391771 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement spring loaded primary check valve assembly, THF, is designed for efficient performance in systems requiring normal phase solvent applications.
  • Designed for use on ACQUITY UPLC, ACQUITY Arc, and Arc HPLC systems.
  • Suitable for systems converted to run normal phase solvents.
  • Type: Valve – Check.
  • Units per package: 1 pk.
  • Recommended annual replacement schedule.
Catalog No. 50-239-1771 Supplier Waters Corp Supplier No. 700011074
This replacement spring loaded primary check valve assembly, THF, is designed for efficient performance in systems requiring normal phase solvent applications.
  • Designed for use on ACQUITY UPLC, ACQUITY Arc, and Arc HPLC systems.
  • Suitable for systems converted to run normal phase solvents.
  • Type: Valve – Check.
  • Units per package: 1 pk.
  • Recommended annual replacement schedule.