Learn More
Chemglass Life Sciences Pressure Relief Valve, Spring Loaded Plug Only
SDP
Catalog No. 50872700
Encompass_Preferred
Pressure Relief Valve, Spring Loaded Plug Only. Replacement part for CG99901 Pressure Relief Valve. Automatic, adjustable pressure relief valve is for use on apparatus where internal pressure build-up is a concern.
Catalog No. 50-872-700 Supplier Chemglass Life Sciences Supplier No. CG99902
Pressure Relief Valve, Spring Loaded Plug Only. Replacement part for CG99901 Pressure Relief Valve. Automatic, adjustable pressure relief valve is for use on apparatus where internal pressure build-up is a concern.