Learn More
Grainger TE-CO Ball Plunger, Stainless Steel, 3/8 in. Body, 5/Pk.
Catalog No. 502785143
Encompass_Preferred
These slotted ball plungers are designed for applications requiring controlled end forces. Featuring a durable stainless steel body and a nylon ball, these components are suitable for various indexing, positioning, and holding tasks in alignment with TCMAI standards.
  • Features a slotted drive design for easy installation with a screwdriver.
  • Constructed with a stainless steel body and a nylon ball.
  • Includes a thread-locking element for secure positioning.
  • Provides a light end force with an initial force of 2.5 lbf and a final force of 5 lbf.
  • Designed for inch measurement systems with 3/8 in.-16 threading.
Catalog No. 50-278-5143 Supplier Grainger Supplier No. 19A723
These slotted ball plungers are designed for applications requiring controlled end forces. Featuring a durable stainless steel body and a nylon ball, these components are suitable for various indexing, positioning, and holding tasks in alignment with TCMAI standards.
  • Features a slotted drive design for easy installation with a screwdriver.
  • Constructed with a stainless steel body and a nylon ball.
  • Includes a thread-locking element for secure positioning.
  • Provides a light end force with an initial force of 2.5 lbf and a final force of 5 lbf.
  • Designed for inch measurement systems with 3/8 in.-16 threading.