Learn More
Grainger TE-CO Ball Plunger, Stainless Steel, 3/8 in Diameter, 3/8 in.-16 Thread, 5/Pk.
Catalog No. 502785580
Encompass_Preferred
These slotted ball plungers are designed for applications requiring controlled end forces. Constructed with a 303 stainless steel body and a nylon ball, these components provide a side-load tolerant solution for precision mechanical assemblies. Each unit includes a thread-locking element to ensure secure positioning during operation.
  • Features a slotted drive for screwdriver installation
  • Provides initial nose force of 6.00 lbf and final nose force of 21.00 lbf
  • Includes thread-locking element for secure mounting
  • Designed to conform to TCMAI standards
  • Side-load tolerant construction
  • Package quantity of 5 units
Catalog No. 50-278-5580 Supplier Grainger Supplier No. 19A725
These slotted ball plungers are designed for applications requiring controlled end forces. Constructed with a 303 stainless steel body and a nylon ball, these components provide a side-load tolerant solution for precision mechanical assemblies. Each unit includes a thread-locking element to ensure secure positioning during operation.
  • Features a slotted drive for screwdriver installation
  • Provides initial nose force of 6.00 lbf and final nose force of 21.00 lbf
  • Includes thread-locking element for secure mounting
  • Designed to conform to TCMAI standards
  • Side-load tolerant construction
  • Package quantity of 5 units