Learn More
Fine Science Tools Vannas Spring Scissors, 2mm Cutting Edge, 8 cm
SDP
Catalog No. NC9391828
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 2mm/ Tip Diameter: 0.05mm/ Alloy / Material: Stainless Steel/ Length: 8cm/ Serrated: No
Catalog No. NC9391828 Supplier Fine Science Tools Supplier No. 1500003
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 2mm/ Tip Diameter: 0.05mm/ Alloy / Material: Stainless Steel/ Length: 8cm/ Serrated: No