Learn More
Fine Science Tools Vannas-Tübingen Spring Scissors, Stainless Steel, 5 mm Cutting Edge, 9.5 cm
SDP
Catalog No. NC0786515
Encompass
Tip Shape: Angled Up/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 9.5cm/ Serrated: No
Catalog No. NC0786515 Supplier Fine Science Tools Supplier No. 1500508
Tip Shape: Angled Up/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 9.5cm/ Serrated: No