Learn More
World Precision Instrument Spring Scissors, Super Cut, 10 cm
SDP
Supplier:  World Precision Instrument WP22201
Encompass
Our Spring Scissors with one serrated blade are surgical scissors designed for making small, precision cuts. They are 10cm long, and fit comfortably into your hand. These spring scissors have one edge that is serrated and one that is sharp. The serrations provide a firm grasp of the tissue when the sharp blade shears the tissue. The end result is a clean cut with minimal damage to surrounding tissues. These have straight or angled tips. The spring mechanism that allows for smooth, precise cutting action, which reduces the risk of tissue damage or trauma. The spring scissors also help to mitigate hand fatigue, because they spring back naturally after you make your cut. The spring handled surgical scissors are preferred by left-handed surgeons, because they can be used ambidextrously. This gives the left-handed surgeon a more natural cutting action, which means the surgeon has greater control of the tip and the cutting movement.
Catalog No. 50-253-8328