Learn More
Fine Science Tools Vannas Spring Scissors - 2.5mm Cutting Edge
SDP
Catalog No. NC9203661
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 2.5mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 8cm/ Serrated: No. These scissors have been designed to maximize visibility under the microscope by eliminating unnecessary bulk in the blades. A short taper makes these scissors ideal for small, precise cuts.
Catalog No. NC9203661 Supplier Fine Science Tools Supplier No. 1500108
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 2.5mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 8cm/ Serrated: No. These scissors have been designed to maximize visibility under the microscope by eliminating unnecessary bulk in the blades. A short taper makes these scissors ideal for small, precise cuts.