Learn More
Julabo USA Inc Set, extra spring clamps for 8970630. Use with SW22/SW23 shaking water baths.
SDP
Catalog No. 507534095
Encompass_Preferred
Bath Accessories, Julabo, Spring clamps set for basic tray 8970630. Set including 12 spring clamps 135 mm and 5 spring clamps 190 mm length. Additional springs. For use with shaking water bath models SW22, SW23.
Catalog No. 50-753-4095 Supplier Julabo USA Inc Supplier No. 8970631
Bath Accessories, Julabo, Spring clamps set for basic tray 8970630. Set including 12 spring clamps 135 mm and 5 spring clamps 190 mm length. Additional springs. For use with shaking water bath models SW22, SW23.