Learn More
MSC Precision Brand Feeler Gage, Spring Steel, 0.020 in. Thickness, 1/2 in. Width, 5 in. Length, 10/Pk.
Catalog No. 5000512153
Encompass
These feeler gages are manufactured from C1095 high carbon spring steel for durability and precision. They are designed for applications requiring the measurement of tight clearances and the setting of close spacing between mechanical components.
  • Constructed from C1095 high carbon spring steel
  • Ground and polished to a thickness tolerance of ±.00055 in.
  • Hardness rating of RC 48-62
  • Clearly etched with decimal and metric thicknesses
  • Ends are rounded and edges are smooth for enhanced safety
  • Includes a 3/16 in. hole at one end for storage and organization
  • Meets FED-GGG-G-17C certification standards
Catalog No. 50-005-12153 Supplier MSC Supplier No. 53488276
These feeler gages are manufactured from C1095 high carbon spring steel for durability and precision. They are designed for applications requiring the measurement of tight clearances and the setting of close spacing between mechanical components.
  • Constructed from C1095 high carbon spring steel
  • Ground and polished to a thickness tolerance of ±.00055 in.
  • Hardness rating of RC 48-62
  • Clearly etched with decimal and metric thicknesses
  • Ends are rounded and edges are smooth for enhanced safety
  • Includes a 3/16 in. hole at one end for storage and organization
  • Meets FED-GGG-G-17C certification standards