Learn More
Instech Spring for Instech mouse tethers, .065in ID, .010in wire, five 12in (30cm) lengths, non-sterile
SDP
Catalog No. 5081192
Encompass
The Instech PS62 is a replacement tether spring designed for mouse infusion tethers, featuring a .065in inner diameter and .010in wire gauge to provide a lightweight, flexible spring suitable for small‑animal tethering systems. Supplied non‑sterile in packs of five, each spring is 12in (30cm) long and used for building or refurbishing dependable mouse infusion tether assemblies.
Features
  • .065in inner diameter for mouse‑sized tether systems
  • .010in wire gauge for lightweight flexibility
  • 12in (30cm) spring length
  • Non‑sterile replacement springs
  • Packaged in sets of five
Catalog No. 50-811-92 Supplier Instech Supplier No. PS62
The Instech PS62 is a replacement tether spring designed for mouse infusion tethers, featuring a .065in inner diameter and .010in wire gauge to provide a lightweight, flexible spring suitable for small‑animal tethering systems. Supplied non‑sterile in packs of five, each spring is 12in (30cm) long and used for building or refurbishing dependable mouse infusion tether assemblies.
Features
  • .065in inner diameter for mouse‑sized tether systems
  • .010in wire gauge for lightweight flexibility
  • 12in (30cm) spring length
  • Non‑sterile replacement springs
  • Packaged in sets of five