Learn More
Instech Spring for Instech rat tethers, .093in ID, .016in wire, five 12in (30cm) lengths, non-sterile.
SDP
Catalog No. 5081193
Encompass
The Instech PS95 is a replacement tether spring for rat infusion tethers, featuring a .093in inner diameter and .016in wire gauge to provide the proper balance of strength and flexibility for standard rat tether assemblies. Each non‑sterile spring measures 12in (30cm) in length and is supplied in packs of five for constructing or refurbishing durable tether systems used in infusion or sampling studies.
Features
  • .093in inner diameter sized for rat tether applications
  • .016in wire gauge for reliable flexibility and durability
  • 12in (30cm) spring length
  • Non‑sterile replacement springs
  • Packaged in sets of five
This item is not returnable. View return policy
Catalog No. 50-811-93 Supplier Instech Supplier No. PS95
 Restricted Item
Add to Cart
Add to Cart
The Instech PS95 is a replacement tether spring for rat infusion tethers, featuring a .093in inner diameter and .016in wire gauge to provide the proper balance of strength and flexibility for standard rat tether assemblies. Each non‑sterile spring measures 12in (30cm) in length and is supplied in packs of five for constructing or refurbishing durable tether systems used in infusion or sampling studies.
Features
  • .093in inner diameter sized for rat tether applications
  • .016in wire gauge for reliable flexibility and durability
  • 12in (30cm) spring length
  • Non‑sterile replacement springs
  • Packaged in sets of five