Learn More
Instech Spring for Instech infusion tethers, .108in ID, .022in wire, five 12in (30cm) lengths, non-sterile.
SDP
Catalog No. 5081191
Encompass
The Instech PS105 is a replacement tether spring designed for use in rodent infusion tethers, featuring a .108in inner diameter and .022in wire gauge to provide strength and flexibility during tethered infusion or sampling studies. Supplied non‑sterile in packs of five, each spring is 12in (30cm) long and suitable for constructing or repairing durable tether assemblies.
Features
  • .108in inner diameter for stable tether construction
  • .022in wire gauge for strength and flexibility
  • 12in (30cm) spring length
  • Non‑sterile replacement springs
  • Packaged in sets of five
Catalog No. 50-811-91 Supplier Instech Supplier No. PS105
The Instech PS105 is a replacement tether spring designed for use in rodent infusion tethers, featuring a .108in inner diameter and .022in wire gauge to provide strength and flexibility during tethered infusion or sampling studies. Supplied non‑sterile in packs of five, each spring is 12in (30cm) long and suitable for constructing or repairing durable tether assemblies.
Features
  • .108in inner diameter for stable tether construction
  • .022in wire gauge for strength and flexibility
  • 12in (30cm) spring length
  • Non‑sterile replacement springs
  • Packaged in sets of five