Learn More
Julabo USA Inc Each, All purpose Spring tray for Erlenmeyer Flasks
SDP
Catalog No. 507534094
Encompass_Preferred
Bath Accessories, Julabo, All purpose Spring tray for Erlenmeyer Flasks from 25mL to 1L, including spring clamps. For use with Shaking Water baths SW22 and SW23.
This item is not returnable. View return policy
Catalog No. 50-753-4094 Supplier Julabo USA Inc Supplier No. 8970630
Bath Accessories, Julabo, All purpose Spring tray for Erlenmeyer Flasks from 25mL to 1L, including spring clamps. For use with Shaking Water baths SW22 and SW23.