Learn More
Hach Company Cap only for 177 mL Demineralizer Bottle
Supplier:  Hach Company 591200
Encompass
Narrow mouth, polyethylene bottle. Contains monobed resin. Flip spout. Great way to deionize small volumes of water. Used with Hach Portable Laboratories.
Catalog No. 50-199-4814