Learn More
Analytical Sales & Services Sprite 300+ C4 Column. 5µm, 20mm x 2.1mm ID
Catalog No. 5000806047
Encompass
Sprite 300+ C4 Column. 5µm, 20mm x 2.1mm ID\n\n• For applications requiring high resolution and selectivity\n• Fast HPLC Analysis\n• LC-MS Applications\n• PEEK / Titanium design
Catalog No. 50-008-06047 Supplier Analytical Sales & Services Supplier No. SPRITE 94202
Sprite 300+ C4 Column. 5µm, 20mm x 2.1mm ID\n\n• For applications requiring high resolution and selectivity\n• Fast HPLC Analysis\n• LC-MS Applications\n• PEEK / Titanium design