Learn More
Analytical Sales & Services Sprite 300+ C8 Column. 5µm, 20mm x 2.1mm ID
Catalog No. 5000805651
Encompass
Sprite 300+ C8 Column. 5µm, 20mm x 2.1mm ID. • For applications requiring high resolution and selectivity • Fast HPLC Analysis • LC-MS Applications • PEEK / Titanium design
Catalog No. 50-008-05651 Supplier Analytical Sales & Services Supplier No. SPRITE 98202
Sprite 300+ C8 Column. 5µm, 20mm x 2.1mm ID. • For applications requiring high resolution and selectivity • Fast HPLC Analysis • LC-MS Applications • PEEK / Titanium design