Learn More
Analytical Sales & Services Sprite Armor C18 Column. 10µm, 20mm x 2.1mm ID
Catalog No. 5000805786
Encompass
Sprite Armor C18 Column. 10µm, 20mm x 2.1mm ID. Monofunctional with unique polar and bulky end-capping.
Catalog No. 50-008-05786 Supplier Analytical Sales & Services Supplier No. SPRITE A101822
Add to Cart
Add to Cart
Sprite Armor C18 Column. 10µm, 20mm x 2.1mm ID. Monofunctional with unique polar and bulky end-capping.