Learn More
Analytical Sales & Services Sprite Armor C18 Column. 10µm, 40mm x 2.1mm ID
Catalog No. 5000804936
Encompass
Sprite Armor C18 Column. 10µm, 40mm x 2.1mm ID. Monofunctional with unique polar and bulky end-capping.
Catalog No. 50-008-04936 Supplier Analytical Sales & Services Supplier No. SPRITE A101842
Sprite Armor C18 Column. 10µm, 40mm x 2.1mm ID. Monofunctional with unique polar and bulky end-capping.