Learn More
Analytical Sales & Services Sprite Armor C18 Column. 5µm, 10mm x 2.1mm ID
Catalog No. 5000806044
Encompass
Sprite Armor C18 Column. 5µm, 10mm x 2.1mm ID. Monofunctional with unique polar and bulky end-capping.
Catalog No. 50-008-06044 Supplier Analytical Sales & Services Supplier No. SPRITE AC1832
Sprite Armor C18 Column. 5µm, 10mm x 2.1mm ID. Monofunctional with unique polar and bulky end-capping.