Learn More
Analytical Sales & Services Sprite Lancer C18 Column. 10um, 40mm x 2.1mm ID
Catalog No. 5000806652
Encompass
Sprite Lancer C18 Column. 10um, 40mm x 2.1mm ID. Extremely well end-capped monofunctional phase.
Catalog No. 50-008-06652 Supplier Analytical Sales & Services Supplier No. SPRITE L101842
Sprite Lancer C18 Column. 10um, 40mm x 2.1mm ID. Extremely well end-capped monofunctional phase.