Learn More
Analytical Sales & Services Sprite Lancer C18 Column. 5µm, 10mm x 2.1mm ID
Catalog No. 5000805653
Encompass
Sprite Lancer C18 Column. 5µm, 10mm x 2.1mm ID. Extremely well end-capped monofunctional phase.
Catalog No. 50-008-05653 Supplier Analytical Sales & Services Supplier No. SPRITE LC1832
Sprite Lancer C18 Column. 5µm, 10mm x 2.1mm ID. Extremely well end-capped monofunctional phase.