Learn More
Analytical Sales & Services Sprite Lancer C18 Column. 5µm, 40mm x 2.1mm ID
Catalog No. 5000805790
Encompass
Sprite Lancer C18 Column. 5µm, 40mm x 2.1mm ID. Extremely well end-capped monofunctional phase.
Catalog No. 50-008-05790 Supplier Analytical Sales & Services Supplier No. SPRITE LC1842
Sprite Lancer C18 Column. 5µm, 40mm x 2.1mm ID. Extremely well end-capped monofunctional phase.