Learn More
Enzo Life Sciences Super Killer TRAIL Protein (soluble) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences ALX201115C010
Encompass_Preferred
Enhanced ligand with improved stability providing significantly enhanced immune activation. Alternative name: TNFSF 10, Apo-2L, CD253. MW: ~26kDa. Endotoxin content: <0.01EU/µg purified protein (LAL test). Purity: ≥98% (SDS-PAGE). MS analysis, <1% impurity. Formulation: Liquid. In 20mM HEPES, pH 7.4, containing 150mM sodium chloride, 0.005% Tween 20, 1% sucrose, 1mM DTT. Source: Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide. The active multimeric conformation is stabilized by an inserted mutation allowing an additional CC-bridge. Specificity: Binds to human TRAIL receptors. Recommended: Prod. No. ALX-505-005, special storage and dilution buffer. UniProt: P50591.
Catalog No. NC9009178