Learn More
Aviva Systems Biology Corp SPRN Antibody - N-terminal region (ARP64847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPRN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human SPRN. Alternate names: FLJ41197, SHADOO, SHO, bA108K14.1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-288-383