Learn More
Chemglass Life Sciences Spring, Stainless Steel, 25mm Coil Length
SDP
Catalog No. 50873710
Encompass_Preferred
Spring, Stainless Steel, 25mm Coil Length. Stainless steel spring used to connect apparatus having hooks. All springs have an enlarged 1/4" ID loop at each end for easy connection to almost any size hook. Diameter of the coils on all sizes is 0.190". Length listed is the length between the loops. SOLD 12/PKG.
Catalog No. 50-873-710 Supplier Chemglass Life Sciences Supplier No. CG11003
Spring, Stainless Steel, 25mm Coil Length. Stainless steel spring used to connect apparatus having hooks. All springs have an enlarged 1/4" ID loop at each end for easy connection to almost any size hook. Diameter of the coils on all sizes is 0.190". Length listed is the length between the loops. SOLD 12/PKG.