Learn More
Chemglass Life Sciences Spring, Stainless Steel, 6.35mm Coil Length
SDP
Catalog No. 50873708
Encompass_Preferred
Stainless steel spring used to connect apparatus having hooks. 6.35mm Coil Length. All springs have an enlarged 1/4" ID loop at each end for easy connection to almost any size hook. Diameter of the coils on all sizes is 0.190". Length listed is the length between the loops. SOLD 12/PKG.
Catalog No. 50-873-708 Supplier Chemglass Life Sciences Supplier No. CG11001
Stainless steel spring used to connect apparatus having hooks. 6.35mm Coil Length. All springs have an enlarged 1/4" ID loop at each end for easy connection to almost any size hook. Diameter of the coils on all sizes is 0.190". Length listed is the length between the loops. SOLD 12/PKG.