Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack with Spring Clips for Standard 2" Boxes, 11 box capacity
Catalog No. 501960696 Shop All Cole-Parmer Products
Encompass_Preferred
Use these freezer racks with a wide variety of chest freezers, and liquid nitrogen tanks. They accommodates standard two-inch cardboard, plastic and metal boxes and come with spring clips to securely hold boxes in place. The racks feature dual stainless steel handles for easy handling and are made of sturdy corrosion-resistant stainless steel.
Catalog No. 50-196-0696 Supplier Cole-Parmer Supplier No. 0439883
Use these freezer racks with a wide variety of chest freezers, and liquid nitrogen tanks. They accommodates standard two-inch cardboard, plastic and metal boxes and come with spring clips to securely hold boxes in place. The racks feature dual stainless steel handles for easy handling and are made of sturdy corrosion-resistant stainless steel.