Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack with Spring Clips for Mini 2" Boxes, 12 box capacity
Catalog No. 501960700 Shop All Cole-Parmer Products
Encompass_Preferred
Use these vertical racks with a wide variety of chest freezers and liquid nitrogen tanks. Included locking rod provides added security that the boxes will stay in place.
Catalog No. 50-196-0700 Supplier Cole-Parmer Supplier No. 0439904
Use these vertical racks with a wide variety of chest freezers and liquid nitrogen tanks. Included locking rod provides added security that the boxes will stay in place.