Learn More
Havels Sprotte needle 20G X 3-1/2" (90MM) With Winged Introducer
SDP
Catalog No. NC1706345
Encompass
The Atraumatic Sprotte Spinal Needle for regional anesthesia and lumbar puncture reduces post-spinal headaches by a factor of ten! The conical point reduces dura trauma because it spreads dura fibers - it doesn't cut. The special opening allows CSF to flow freely and the rounded edges prevent coring.
This item is not returnable. View return policy
Catalog No. NC1706345 Supplier Havels Supplier No. WINS2090
The Atraumatic Sprotte Spinal Needle for regional anesthesia and lumbar puncture reduces post-spinal headaches by a factor of ten! The conical point reduces dura trauma because it spreads dura fibers - it doesn't cut. The special opening allows CSF to flow freely and the rounded edges prevent coring.