Learn More
Electron Microscopy Sciences Sprout Sprout For UK, 220/230 VAC
SDP
Catalog No. 5027210
Encompass_Preferred
Newly improved rubber footplate includes soft feet to increase stability and deadens vibration. Ideal for quick spin-downs from tube walls and caps. Use to microfilter cell separations and HPLC samples. Most spin last around 20 seconds, so Sprout is designed for easy repeat opening. Ergonomically designed lid fits the palm of the hand comfortably.
Operation is very simple: load tubes, close lid, press ON/OFFF button. For repeated spin, simply open or close the lid to operate. Internal safety switch ensure unit will not operate with lid open.
This personal centrifuge uses minimal benchtop space. Removable cord for easy storage. Unit is ETL, cETL, and CE marked and also complies with European RoHS directive.
Maximum Speed : 600 rpm. Maximum RCF: 2000 x g.
Catalog No. 50-272-10 Supplier Electron Microscopy Sciences Supplier No. 72360UK
Newly improved rubber footplate includes soft feet to increase stability and deadens vibration. Ideal for quick spin-downs from tube walls and caps. Use to microfilter cell separations and HPLC samples. Most spin last around 20 seconds, so Sprout is designed for easy repeat opening. Ergonomically designed lid fits the palm of the hand comfortably.
Operation is very simple: load tubes, close lid, press ON/OFFF button. For repeated spin, simply open or close the lid to operate. Internal safety switch ensure unit will not operate with lid open.
This personal centrifuge uses minimal benchtop space. Removable cord for easy storage. Unit is ETL, cETL, and CE marked and also complies with European RoHS directive.
Maximum Speed : 600 rpm. Maximum RCF: 2000 x g.