Learn More
Electron Microscopy Sciences Plus Mini Centrifuge, 1/EA
SDP
Catalog No. 502091016
Encompass_Preferred
With an ergonomic design, the Sprout Plus Mini Centrifuge brings increased efficiency to the lab. Tool-free SnapSpin rotors and quick-spin options make the centrifuges fast and easy to use. The lid fits in the palm of the hand for easy repeat opening and quick tube extraction. A weighted rubberized base eliminates potential movement and dampens vibration and noise. The Sprout Plus is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1.5/2.0, 0.5, 0.2 ml) for customized use. The centrifuge housing is made from ABS plastic and lids are polycarbonate plastic for easy cleaning. The Sprout Plus has the same footprint as the original Sprout, with added features of locking lid for added protection, larger lid to accommodate longer and skirted tubes, as well as taller tube lids, and breaking function to stop the unit when powered off or lid open. Color: Blue.
Catalog No. 50-209-1016 Supplier Electron Microscopy Sciences Supplier No. 7236201
With an ergonomic design, the Sprout Plus Mini Centrifuge brings increased efficiency to the lab. Tool-free SnapSpin rotors and quick-spin options make the centrifuges fast and easy to use. The lid fits in the palm of the hand for easy repeat opening and quick tube extraction. A weighted rubberized base eliminates potential movement and dampens vibration and noise. The Sprout Plus is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1.5/2.0, 0.5, 0.2 ml) for customized use. The centrifuge housing is made from ABS plastic and lids are polycarbonate plastic for easy cleaning. The Sprout Plus has the same footprint as the original Sprout, with added features of locking lid for added protection, larger lid to accommodate longer and skirted tubes, as well as taller tube lids, and breaking function to stop the unit when powered off or lid open. Color: Blue.