Learn More
Research Products International Corp SPROUT PLUS MINI-CENTRIFUGE BL
SDP
Catalog No. NC2014421
Encompass
Sprout Plus Mini-Centrifuge is ideal for quick spinning of samples at the workbench, in a cold room, or fume hood. New locking lid fits in the palm of the hand for easy, repeat opening and one hand tube extraction. Ergonomic lid design accommodates longer and skirted tubes and tall tube lids. Supplied with convenient tool-free Snapspin rotors that quickly snap into unit and hold 6 x 1.5-2.0ml tubes or 16 x 0.2ml PCR tubes (single or 2x8 strip); 6 x 0.2ml tube adapters; 6 x 0.5ml tube adapters; 4 Lid O-rings; storage case for rotors and adapters; tube rack; and low voltage, double insulated power adapter with 4 interchangeable plugs for power adapter. Features include fast braking motor that stops spinning when lid is opened; maintenance free motor; and weighted rubberized base that eliminates movement and dampens vibration and noise.
Catalog No. NC2014421 Supplier Research Products International Corp Supplier No. 400802B
Sprout Plus Mini-Centrifuge is ideal for quick spinning of samples at the workbench, in a cold room, or fume hood. New locking lid fits in the palm of the hand for easy, repeat opening and one hand tube extraction. Ergonomic lid design accommodates longer and skirted tubes and tall tube lids. Supplied with convenient tool-free Snapspin rotors that quickly snap into unit and hold 6 x 1.5-2.0ml tubes or 16 x 0.2ml PCR tubes (single or 2x8 strip); 6 x 0.2ml tube adapters; 6 x 0.5ml tube adapters; 4 Lid O-rings; storage case for rotors and adapters; tube rack; and low voltage, double insulated power adapter with 4 interchangeable plugs for power adapter. Features include fast braking motor that stops spinning when lid is opened; maintenance free motor; and weighted rubberized base that eliminates movement and dampens vibration and noise.