Learn More
Aviva Systems Biology Corp SPRR1A Antibody - middle region (ARP63528_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63528_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPRR1A. It was validated on Western Blot. . Alternate names: SPRKApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-998