Learn More
Aviva Systems Biology Corp SPRR3 Antibody - middle region (ARP42236_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42236_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPRR3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPRR3. Alternate names: -ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58157