Learn More
Aviva Systems Biology Corp SPRY1 Antibody - N-terminal region (ARP50521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPRY1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human SPRY1. Alternate names: hSPRY1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54069