Learn More
Aviva Systems Biology Corp SPRY3 Antibody - N-terminal region (ARP50520_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50520_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPRY3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SPRY3. Alternate names: HSPRY3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-516