Learn More
Aviva Systems Biology Corp SPRYD3 Antibody - N-terminal region (ARP60687_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60687_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPRYD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SPRYD3. Alternate names: FLJ14800ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-125