Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PRO-SOURCE Cleaner and Degreaser, Butyl-Free, Fruit Scent, 1 qt

Catalog No. 502870583
Encompass

This all-purpose cleaner and degreaser is formulated for industrial use on hard surfaces. It is a butyl-free, liquid solution with a fruity scent, designed for effective cleaning.

  • Formulated for industrial use
  • Effective on hard surfaces
  • Butyl-free formula
  • EcoLogo certified
  • Biodegradable
  • Light red, clear liquid appearance
  • Ph 10.0-10.5
  • Specific gravity 1.01

Catalog No. 50-287-0583 Supplier MSC Supplier No. 55049670
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This all-purpose cleaner and degreaser is formulated for industrial use on hard surfaces. It is a butyl-free, liquid solution with a fruity scent, designed for effective cleaning.

  • Formulated for industrial use
  • Effective on hard surfaces
  • Butyl-free formula
  • EcoLogo certified
  • Biodegradable
  • Light red, clear liquid appearance
  • Ph 10.0-10.5
  • Specific gravity 1.01

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.