Learn More
Aviva Systems Biology Corp SPSB2 Antibody - N-terminal region (ARP60607_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPSB2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SPSB2. Alternate names: FLJ17395, GRCC9, MGC2519, SSB2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-100