Learn More
Accuflow Systems Saint-Gobain Peristaltic Pump Tubing, Platinum-Cured Silicone, 0.047 in ID
Catalog No. NC2389611
Encompass_Preferred
Peristaltic pump tubing engineered for high-accuracy pharmaceutical and bioprocess dispensing. Platinum-cured silicone formulation minimizes spallation and delivers consistent filling performance and extended pump life in final-fill and precision-pumping applications.
  • Platinum-cured silicone for low spallation and bioprocess compatibility.
  • 0.047 in (1.2 mm) inner diameter, 0.172 in (4.4 mm) outer diameter, 0.063 in (1.6 mm) wall.
  • 60 Shore A nominal hardness.
  • Typical pump life greater than 250 hours under standard operating conditions.
  • Autoclavable; gamma irradiation up to 5.0 Mrad (50 kGy); ethylene oxide compatible.
  • Burst pressure approximately 174 psi (12.0 bar).
  • Compliance and test support: USP <88> Class VI, USP <87>, ISO 10993-5, ADCF/BSE/TSE statements; full extractables report per BPOG available.
Catalog No. NC2389611 Supplier Accuflow Systems Supplier No. SPT60L0472
Peristaltic pump tubing engineered for high-accuracy pharmaceutical and bioprocess dispensing. Platinum-cured silicone formulation minimizes spallation and delivers consistent filling performance and extended pump life in final-fill and precision-pumping applications.
  • Platinum-cured silicone for low spallation and bioprocess compatibility.
  • 0.047 in (1.2 mm) inner diameter, 0.172 in (4.4 mm) outer diameter, 0.063 in (1.6 mm) wall.
  • 60 Shore A nominal hardness.
  • Typical pump life greater than 250 hours under standard operating conditions.
  • Autoclavable; gamma irradiation up to 5.0 Mrad (50 kGy); ethylene oxide compatible.
  • Burst pressure approximately 174 psi (12.0 bar).
  • Compliance and test support: USP <88> Class VI, USP <87>, ISO 10993-5, ADCF/BSE/TSE statements; full extractables report per BPOG available.