Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, For 24/40-24/25 Outer Joints, White
SDP
Catalog No. CG302228
Encompass_Preferred
Septum Stopper, Sleeve Type, For 24/40-24/25 Outer Joints, White. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. CG302228 Supplier Chemglass Life Sciences Supplier No. CG302228
Septum Stopper, Sleeve Type, For 24/40-24/25 Outer Joints, White. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.