Learn More
Aviva Systems Biology Corp SPTAN1 Antibody - N-terminal region (ARP58532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPTAN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPTAN1. Alternate names: (ALPHA)II-SPECTRIN, FLJ44613, NEAS, EIEE5, SPTA2ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62347