Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6712MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: SPTBN2/1583. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HeLa cells. Pancreas or Liver. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human SPTBN2 fragment (aa356-475). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 246kDa.Chromosome: 11q13.2. Gene Symbol: SPTBN2. Gene ID: 6712. Ensemble: ENSG00000173898. Uniprot: XP_016873664.
Catalog No. 50-188-5191