Learn More
Cayman Chemical SpThymIdIn5O 1thIotrIp 2.5mg
SDP
Catalog No. 502918989
Encompass_Preferred
An isomer of TTP-α-S; binds to HIV-1 reverse transcriptase (Kds = 2.88, 4.23, and 20.21 µM in the presence of magnesium, manganate, and cobalt, respectively); inhibits deoxynucleoside triphosphate triphosphohydrolase SAMHD1 (Ki = 46 nM)
Catalog No. 50-291-8989 Supplier Cayman Chemical Supplier No. 386902.5 MG
An isomer of TTP-α-S; binds to HIV-1 reverse transcriptase (Kds = 2.88, 4.23, and 20.21 µM in the presence of magnesium, manganate, and cobalt, respectively); inhibits deoxynucleoside triphosphate triphosphohydrolase SAMHD1 (Ki = 46 nM)