Learn More
Aviva Systems Biology Corp SPTLC1 Antibody - middle region (ARP46721_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46721_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPTLC1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SPTLC1. Alternate names: HSAN, HSAN1, HSN1, LBC1, LCB1, MGC14645, SPT1, SPTI, hLCB1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53487