Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, For 29/26-29/42 Outer Joints, Red
SDP
Supplier:  Chemglass Life Sciences CG302288
Encompass_Preferred
Solid Rubber Stopper, #00, Black. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250°F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (15mm), Bottom (10mm). Length: (25mm).
Catalog No. 50-872-546