Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, For 14/20-14/35 Outer Joints, Red
SDP
Catalog No. CG302206
Encompass_Preferred
Septum Stopper, Sleeve Type, For 14/20-14/35 Outer Joints, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn. Available in either RED or WHITE rubber and sold in the package quantities listed below.
This item is not returnable. View return policy
Catalog No. CG302206 Supplier Chemglass Life Sciences Supplier No. CG302206
Septum Stopper, Sleeve Type, For 14/20-14/35 Outer Joints, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn. Available in either RED or WHITE rubber and sold in the package quantities listed below.