Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, for 8mm OD Standard Wall or 9mm OD Medium Wall, Red
SDP
Catalog No. CG302205
Encompass_Preferred
Septum Stopper, Sleeve Type, for 8mm OD Standard Wall or 9mm OD Medium Wall, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. CG302205 Supplier Chemglass Life Sciences Supplier No. CG302205
Septum Stopper, Sleeve Type, for 8mm OD Standard Wall or 9mm OD Medium Wall, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.