Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, For 19/22-19/38 Outer Joints, White
SDP
Catalog No. CG302226
Encompass_Preferred
Septum Stopper, Sleeve Type, For 19/22-19/38 Outer Joints, White. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. CG302226 Supplier Chemglass Life Sciences Supplier No. CG302226
Septum Stopper, Sleeve Type, For 19/22-19/38 Outer Joints, White. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.